Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,3,6-Trifluoroanisole ≥97%

Supplier: Alfa Aesar
Warning

Purity: 97%
CAS No.: 4920-34-7
Molecular Formula: C7H5F3O
Formula Weight: 162.11
Flash Point: 45°(113°F)
Refractive Index: 1.4450
Boiling Point: 146°
Beilstein Reference: 2557137
UN No.: 3271
MDL No.: MFCD01631501


Formula: C₇H₅F₃O
MW: 162,11 g/mol
Boiling Pt: 146 °C
Flash Pt: 45 °C (113 °F)
Storage Temperature: Ambient
MDL Number: MFCD01631501
CAS Number: 4920-34-7
EINECS: 000-000-0
UN: 3271
ADR: 3,III

 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 1 g ALFAB24302.03
Supplier Article Number:: B24302.03
Glass bottle for solids 5 g ALFAB24302.06
Supplier Article Number:: B24302.06
Expand All / Collapse All

SPECIFICATIONS

Formula weight 162.11
CAS No. 4920-34-7
Flash point 45°(113°F)
Molecular formula C7H5F3O
UN No. 3271
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping