2-Bromo-4-methylbenzaldehyde 95%
Supplier: Thermo Scientific
|
Warning
|
ALFAH27558.03EA
160
SGD
ALFAH27558.03
2-Bromo-4-methylbenzaldehyde 95%
2-Bromo-4-methylbenzaldehyde
Formula:
C₈H₇BrO MW: 199,05 g/mol Melting Pt: 33…37 °C Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD04039890 CAS Number: 824-54-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...