ALFAA10721

ALFAA10721.14EA 242 SGD
ALFAA10721.14 ALFAA10721.22 ALFAA10721.06
ALFAA10721
Purity: 98+%
CAS No.: 51437-00-4
Molecular Formula: C7H6BrF
Formula Weight: 189.03
Flash Point: 73°(163°F)
Refractive Index: 1.5290
Boiling Point: 94-95°/50mm
Beilstein Reference: 2242693
MDL No.: MFCD00000343
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR