4-Fluoro-α,α,α-trichlorotoluene ≥98%
Supplier: Thermo Scientific
|
Danger
|
ALFAL04560.06EA
134
SGD
ALFAL04560.06
ALFAL04560.14
4-Fluoro-α,α,α-trichlorotoluene ≥98%
4-Fluoro-α,α,α-trichlorotoluene
Purity: 98%
CAS No.: 402-42-6
Molecular Formula: C7H4Cl3F
Formula Weight: 213.47
Refractive Index: 1.5340
Beilstein Reference: 2208417
UN No.: 3265
MDL No.: MFCD00018822
Formula:
C₇H₄Cl₃F MW: 213,47 g/mol Boiling Pt: 212…213 °C (742 mmHg) Density: 1,4385 g/cm³ (30 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00018822 CAS Number: 402-42-6 EINECS: 206-942-0 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...