Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Viscosity baths

Supplier: VWR Collection
VWR®
Viscosity baths for use with popular capillary viscometers are configured to accommodate specific testing and quality control needs, including ASTM D-445. Choice of models with round or square top openings. These transparent polycarbonate baths, with user friendly SD temperature controller, feature four control buttons and on-screen prompts in English. Controller bridge, made from chemical resistant DuraTop™, is easily removed to clean the tank.

  • Large EasyView™ LCD display with alarm or fault icons and prompts plus menus in four languages: French, German, Spanish and English
  • Swivel 180™ rotating controllers, LidDock™ lid stowing system and DuraTop™ chemically resistant deck
  • User-adjustable high and low temperature limits and low liquid level alarms
  • Two speed pressure pump with tap water cooling coil
  • Single point calibration

Maximum pump capacities:
10,2 l/min, 200 mbar

Certifications: Comply with DIN 12876-1, Safety Class I




 

ORDER

Expand All / Collapse All
Description Pk VWR Catalog Number Quantity
Viscosity bath with 5 round holes for viscometers, SD temperature controller 1 VWRI462-0209
Viscosity bath with 3 square holes for viscometers, SD temperature controller 1 VWRI462-0210

SPECIFICATIONS

Model Square holes Round holes
Temperature range (°C) Ambient +10...85
Temperature stability (°C) ±0,04
Display resolution ±0,01
Viscometer positions 3 5
Bath material Polycarbonate
Bath capacity (l) 29
Bath depth (mm) 286
W×D×H (mm) 229×544×526
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty