Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nitrocyclopentane ≥99%

Supplier: Alfa Aesar
Warning

Purity: 99%
CAS No.: 2562-38-1
Molecular Formula: C5H9NO2
Formula Weight: 115.13
Flash Point: 67°(152°F)
Refractive Index: 1.4540
Boiling Point: 179-180°
Beilstein Reference: 907079
MDL No.: MFCD00001362

Formula: C₅H₉NO₂
MW: 115,13 g/mol
Boiling Pt: 40 °C (0,01 -rr)
Density: 1,0776 g/cm³ (23 °C)
CAS Number: 2562-38-1
EINECS: 219-884-6


Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 25 g ALFAL08029.14
Supplier Article Number:: L08029.14
Glass bottle for solids 1 g ALFAL08029.03
Supplier Article Number:: L08029.03
Glass bottle for solids 5 g ALFAL08029.06
Supplier Article Number:: L08029.06
Expand All / Collapse All

SPECIFICATIONS

Formula Weight 115.13
CAS No. 2562-38-1
Purity 99%
Refractive Index 1.4540
Flash Point 67°(152°F)
Boiling Point 179-180°
Molecular Formula C5H9NO2
MDL No. MFCD00001362
Beilstein Reference 907079
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty