Butylcyclohexane ≥99%
Supplier: Thermo Scientific
|
Warning
|
ALFAL08845.18EA
238
SGD
ALFAL08845.18
ALFAL08845.09
Butylcyclohexane ≥99%
Butylcyclohexane
Purity: 99%
CAS No.: 1678-93-9
Molecular Formula: C10H20
Formula Weight: 140.27
Flash Point: 50°(122°F)
Refractive Index: 1.4410
Melting Point: -78°
Boiling Point: 178-180°
Beilstein Reference: 1900631
UN No.: 3295
MDL No.: MFCD00001530
Formula:
C₁₀H₂₀ MW: 140,27 g/mol Boiling Pt: 178…180 °C Melting Pt: –78 °C Density: 0,818 Flash Pt: 50 °C (122 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001530 CAS Number: 1678-93-9 EINECS: 216-837-1 UN: 3295 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...