Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cobalt selenide ≥99% (metals basis)

Supplier: Alfa Aesar
Danger

Purity: 99+%
CAS No.: 1307-99-9
Molecular Formula: CoSe
Formula Weight: 137.89
Physical Form: Powder
Density: 7.650
Solubility: Soluble in HNO3, aqua regia
UN No.: 3283
MDL No.: MFCD00016020


Formula: CoSe
MW: 137,89 g/mol
Melting Pt: 1055 °C
Density: 7,65 g/cm³ (20 °C)
MDL Number: MFCD00016020
CAS Number: 1307-99-9
EINECS: 215-155-1
UN: 3283
ADR: 6.1,III


 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 50 g ALFA40224.18
Supplier Article Number:: 40224.18
Glass bottle for solids 250 g ALFA40224.30
Supplier Article Number:: 40224.30
Expand All / Collapse All

SPECIFICATIONS

Density 7.650
Formula weight 137.89
CAS No. 1307-99-9
Purity 99+%
Physical form Powder
Molecular formula CoSe
MDL No. MFCD00016020
UN No. 3283
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty