Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nickel selenide ≥99.9% (metals basis)

Supplier: Alfa Aesar

Purity: 99.9%
CAS No.: 1314-05-2
Molecular Formula: NiSe
Formula Weight: 137.67
Physical Form: 6mm & Down Pieces
Density: 8.46
Solubility: Insoluble in water. Soluble in aqua regia, HNO3
UN No.: 3077
MDL No.: MFCD00049497

Formula: NiSe
MW: 137,65 g/mol
Melting Pt: 980 °C
Density: 7,217 g/cm³ (25 °C)
MDL Number: MFCD00049497
CAS Number: 1314-05-2
EINECS: 215-216-2
UN: 3077
ADR: 9,III

Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 5 g ALFA13106.06
Supplier Article Number:: 13106.06
Glass bottle for solids 25 g ALFA13106.14
Supplier Article Number:: 13106.14
Expand All / Collapse All

SPECIFICATIONS

Density 8.46
Formula Weight 137.67
CAS No. 1314-05-2
Purity 99.9%
Solubility Insoluble in water. Soluble in aqua regia, HNO3
Physical Form 6mm & Down Pieces
Odor/Scent Odourless
Molecular Formula NiSe
MDL No. MFCD00049497
UN No. 3077
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty