Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4,4-Dimethyl-2-phenyl-2-oxazoline ≥97%

Supplier: Alfa Aesar

Purity: 97%
CAS No.: 19312-06-2
Molecular Formula: C11H13NO
Formula Weight: 175.23
Flash Point: 102°(215°F)
Refractive Index: 1.5320
Melting Point: 20-24°
Boiling Point: 124°/20mm
Beilstein Reference: 132225
MDL No.: MFCD00030446


Formula: C₁₁H₁₃NO
MW: 175,23 g/mol
CAS Number: 19312-06-2

Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 500 g ALFAB25058.36
Supplier Article Number:: B25058.36
Glass bottle for solids 100 g ALFAB25058.22
Supplier Article Number:: B25058.22
Expand All / Collapse All

SPECIFICATIONS

Melting point 20-24°
Formula weight 175.23
CAS No. 19312-06-2
Purity 97%
Refractive index 1.5320
Flash point 102°(215°F)
Boiling point 124°/20mm
Molecular formula C11H13NO
MDL No. MFCD00030446
Beilstein reference 132225
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty