Digital thermo-hygrometer, Traceable® jumbo
VWRI628-0031EA 145 SGD
VWRI628-0031
Digital thermo-hygrometer, Traceable® jumbo
This thermo-hygrometer continuously and simultaneously displays ambient relative humidity and temperature. Rapid response in less than 30 seconds for monitoring conditions overnight, at the weekend, or any other time period.
  • Easy to read jumbo display (25 mm)
  • Probe and cable perform accurately under water
  • Minimum/maximum memories displayed or cleared at any time with the touch of a button
Internal sensor and external probe with cable (3 metres).
Certifications: Compliant with ISO/IEC 17025, ANSI/NCSL Z540-1 and 9001.
Delivery information: Supplied with flip-open stand, wall mount, AAA batteries and Traceable® certificate of calibration.