Yttrium standard solution, 1,000 mg/l Y in dil. nitric acid (from Y2O3) standard for ICP
PANR765915.1208EA 742 SGD
PANR765915.1208
Yttrium standard solution, 1,000 mg/l Y in dil. nitric acid (from Y2O3) standard for ICP
Traceable to NIST