Iridium standard solution, 1000 mg/L Ir in 20% hydrochloric acid (from IrCl₃) standard for ICP
Supplier: PanReac AppliChem
PANR765941.1208EA 725 SGD
PANR765941.1208
Iridium standard solution, 1000 mg/L Ir in 20% hydrochloric acid (from IrCl₃) standard for ICP
Standards Cationic Standards Iridium Standards
Certifications: Traceable to NIST