Ampicillin sodium salt USP

Danger

Synonyms: D(-)-α-Aminobenzylpenicillin Sodium Salt

VWRC0339-25GEA 60.3 SGD
VWRC0339-25G VWRC0339-100G
Ampicillin sodium salt USP
Ampicillin sodium salt
Formula: C₁₆H₁₈N₃NaO₄S
MW: 371,39 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00064313
CAS Number: 69-52-3
EINECS: 200-708-1

Order Now

Specification Test Results

Crystallinity PASS
Dimethylaniline (P/F) PASS
Expiration Date REPORT
Identification PASS
Methylene Chloride 0.2 %
pH (1%, Water) @25C 8.0 - 10.0
Potency (anhydrous) 845 - 988 mcg/mg
Potency (as is) REPORT
Water (KF) 2.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR