Kanamycin sulphate USP
|
Danger
|
Synonyms:
Kanamycin Monosulphate
, Kanamycin A
VWRC0408-25GEA
128.5
SGD
VWRC0408-25G
VWRC0408-1KG
VWRC0408-10G
VWRC0408-100G
Kanamycin sulphate USP
Kanamycin sulphate
Binds to the 70S subunit of bacterial ribosome. Recommended working concentration: 30 μg/ml
Formula:
C₁₈H₃₈N₄O₁₅S MW: 582,6 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00070253 CAS Number: 25389-94-0 EINECS: 246-933-9 |
Specification Test Results
Chromatographic Purity | PASS |
Crystallinity | PASS |
Expiration Date | REPORT |
Identification | PASS |
Loss on Drying | 4.0 % |
pH (1%, Water) @25C | 6.5 - 8.5 |
Potency (Anhydrous) | 750 mcg/mg |
Potency (As is) | REPORT |
Residue on Ignition | 1.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...