Would you like to log in again?
Noise level indicator NI-100
Supplier: 3M
3M
The NI-100 Indicator alerts users to potential harmful noise levels by helping to identify places where any hearing protection products should be worn.
The indicator clips easily to a lapel or shirt pocket. The LED light indicates when a noise level is exceeded, which could be harmful. Red light on LED lamp means that the noise level is more than 85 dB (A) and ear protection must be worn. Green light means that the noise level is below 85 dB (A), hearing protection should be available and can be worn if necessary.
  • Lightweight, simple, stylish design - suitable or many industry sectors
  • Rechargeable battery provides up to 200 hours of use between charges
  • Optional USB cable for recharging
  • The device will power off after approximately 10 hours
The indicator clips easily to a lapel or shirt pocket. The LED light indicates when a noise level exceeded that which could be harmful. Red light on LED lamp means that the noise level is more than 85 dB (A) and ear protection must be worn. Green light means that the noise level is below 85 dB (A), hearing protection should be available and can be worn if necessary.
  • Lightweight, simple stylish design - suitable or many industry sectors
  • Rechargeable battery provides up to 200 hours of use between charges
  • Optional USB cable for recharging
  • The device will power off after approximately 10 hours
Weight: 17 g
W×D×H: 36×13×51 mm
ORDER
Expand All / Collapse All
Description Pk VWR Catalog Number Quantity
NI-100EU Noise Indicator 1 MMMANI100
Supplier Article Number:: NI100
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty