ULT upright freezers, ExF series, Revco®
Supplier: Thermo Scientific
REVCEXF32086SSVEA 0 SGD
REVCEXF32086A REVCEXF32086SSV
ULT upright freezers, ExF series, Revco®
Revco® ExF series ultra low temperature freezers are designed for daily sample protection and dependability.
  • Eye-level, centralized information center, including microprocessor control and monitoring system, ensures that all controls and displays are easy to reach and read
  • Five interior compartment doors to reduce cold air loss, improve temperature recovery after door openings
  • Heavy gauge, cold-rolled steel exterior construction with a powder coat paint finish that resists chipping and rust and 127 mm foamed-in-place,polyurethane insulation
  • Power Management System with low voltage surge protection and buck/boost
  • Access port: 2 (25 mm)
Construction: Exterior: Steel, white coated and interior in Stainless steel.
Temperature range: −50 °C to −86 °C
Ordering information: Optional CO2 or LN2 safety back-up system is available for additional protection in the event of a power or mechanical failure. Optional, built-in chart recorder or data logger for continuous monitoring can be ordered separately, for details of the range of accessories, please ontact your local VWR sales office.