Climatic chambers, KBF, KBF P and KBF LQC series
Supplier: Binder
BIND9020-0327EA 0 SGD
BIND9020-0327 BIND9020-0325 BIND9020-0323 BIND9020-0321
Climatic chambers, KBF, KBF P and KBF LQC series
Testing Chambers Climatic Chambers
The KBF series is ideal for reliable stability tests and precise maintenance of constant climatic conditions. Constant temperature and humidity values are the outstanding features of this range. These constant climate chambers have a large reserve capacity and a variety of optional features. They also comply with all applicable guidelines with respect to programs and documentation requirements, such as ICH, FDA, GMP and GLP. Units can be operated with tap water with BINDER PURE AQUA SERVICE, a flexible water purification system (optional) or demineralised, deionised, distilled, ultra pure water.
  • Electronically controlled humidification and dehumidification system with capacitive humidity sensor
  • Environmentally friendly refrigerant R134a
  • Suitable for stability tests according to ICH guideline Q1A (R2)
  • Illumination is specified for stability in accordance with ICH guideline Q1B, option 2
  • Electronically controlled APT.line™ preheating chamber and refrigeration system gives temperature accuracy and reproducible results
Units have an inner glass door with smooth inner face and seals, access port with Ø 30 mm silicone plug on the left. MCS controller with colour LCD screen with 25 storable programs each with 100 sections for max. 500 program segments. Integrated electronic chart recorder, variety of options for the graphic display of process parameters and a real time clock. Microprocessor controlled humidification and dehumidification system with a humidity range of 10 - 80% RH, humidity accuracy levels of ±1,5% RH. All models have an independent adjustable temperature safety device Class 3.1 with a visual alarm, which saves the samples and the incubator. Complete safety connection kit for water supply and drainage.
KBF series - for precise simulation of constant climatic conditions.
KBF P series - with ICH-compliant illumination, the one source solution for photostability tests.
KBF LQC series - with ICH-compliant illumination and patented light measurement, Light Quantum Control (LQC).
Delivery information: Supplied with two steel racks (KBF P 720 and KBF LQC 720 have three) and Binder test certificate, Ethernet interface for communication software (APT-COM™ DataControl-System software optional). Castors on units of 240 litres or more. RS422 interface available as an option. Calibration and validation is possible, for details please contact your local Avantor sales office.