Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Ethyl-3-nonanol erythro + threo ≥99%

Supplier: Alfa Aesar

Purity: 99%
CAS No.: 19780-71-3
Molecular Formula: C11H24O
Formula Weight: 172.31
MDL No.: MFCD00046677


Formula: C₁₁H₂₄O
CAS Number: 19780-71-3


 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 1 g ALFAB20403.03
Supplier Article Number:: B20403.03
Glass bottle for solids 25 g ALFAB20403.14
Supplier Article Number:: B20403.14
Glass bottle for solids 5 g ALFAB20403.06
Supplier Article Number:: B20403.06
Expand All / Collapse All

SPECIFICATIONS

Formula weight 172.31
CAS No. 19780-71-3
Purity 99%
Molecular formula C11H24O
MDL No. MFCD00046677
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty