Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,6-Dichloro-3-(trifluoromethyl)benzyl bromide 97%, JRD

Supplier: Alfa Aesar
Danger



Formula: C₈H₄BrCl₂F₃
MW: 307,93
Melting Pt: 69…72 °C
Storage Temperature: Ambient
MDL Number: MFCD11519349
CAS Number: 1092461-07-8
UN: 3261
ADR: 8,II

 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 5 g ALFAH31792.06
Supplier Article Number:: H31792.06
Glass bottle for solids 1 g ALFAH31792.03
Supplier Article Number:: H31792.03
Expand All / Collapse All

SPECIFICATIONS

Melting point 69-72°
Formula weight 307.93
Purity 97%
Molecular formula C8H4BrCl2F3
MDL No. MFCD11519349
UN No. 3261
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty