Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ß-Nicotinamide adenine dinucleotide phosphate (NADP, oxidized form) trihydrate ≥97%, reagent grade

Supplier: VWR Chemicals

Formula: C₂₁H₂₈N₇O₁₇P₃·3H₂O
Storage Temperature: Freezer
MDL Number: MFCD10567218
CAS Number: 53-59-8
EINECS: 200-178-1
UN: 0000
Specification Test Results
Heavy Metals 0.002 %
Moisture (KF) REPORT
Purity (Anhydrous) 97 %
Sodium 5.0 %

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle 1 g VWRC0760-1G
Glass bottle 5 g VWRC0760-5G
Glass bottle 500 mg VWRC0760-500MG
Expand All / Collapse All
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty