Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Chloro-2,3,4-trifluorobenzene ≥99%

Supplier: Alfa Aesar

Purity: 99+%
CAS No.: 36556-42-0
Molecular Formula: C6H2ClF3
Formula Weight: 166.53
MDL No.: MFCD00042222


Formula: C₆H₂ClF₃
CAS Number: 36556-42-0


 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 25 g ALFAB23060.14
Supplier Article Number:: B23060.14
Glass bottle for solids 5 g ALFAB23060.06
Supplier Article Number:: B23060.06
Expand All / Collapse All

SPECIFICATIONS

Formula weight 166.53
CAS No. 36556-42-0
Purity 99+%
Molecular formula C6H2ClF3
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty