5-Chloro-2-(trifluoromethoxy)benzyl bromide 97%, JRD

Danger

ALFAH32620.03EA 339 SGD
ALFAH32620.03 ALFAH32620.MD
5-Chloro-2-(trifluoromethoxy)benzyl bromide 97%, JRD
5-Chloro-2-(trifluoromethoxy)benzyl bromide
Formula: C₈H₅BrClF₃O
MW: 289,48 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11519358
CAS Number: 1092461-19-2
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR