Vancomycin hydrochloride USP

Synonyms: Vancomycin supplement

VWRC0990-5GEA 118.95 SGD
VWRC0990-5G VWRC0990-250MG VWRC0990-1G VWRC0990-100MG
Vancomycin hydrochloride USP
Vancomycin hydrochloride
Formula: C₆₆H₇₅Cl₂N₉O₂₄·HCl
MW: 1485,71 g/mol
MDL Number: MFCD03613611
CAS Number: 1404-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR