2-Amino-3-bromo-5-nitropyridine 98%

Warning

Synonyms: 3-Bromo-5-nitro-2-pyridylamine3-Bromo-5-nitropyridin-2-amine

ALFAH33792.06EA 47.4 SGD
ALFAH33792.06 ALFAH33792.14
2-Amino-3-bromo-5-nitropyridine 98%
2-Amino-3-bromo-5-nitropyridine
Formula: C₅H₄BrN₃O₂
MW: 218,01 g/mol
Melting Pt: 218…222 °C
Storage Temperature: Ambient
MDL Number: MFCD00955628
CAS Number: 15862-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR