Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(4-Methoxycarbonylbenzyl)triphenylphosphonium bromide ≥98%

Supplier: Alfa Aesar
Warning



Formula: C₂₇H₂₄BrO₂P
MW: 491,37
Melting Pt: 252…258 °C
Storage Temperature: Ambient
MDL Number: MFCD00082765
CAS Number: 1253-46-9

 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 10 g ALFAL09423.09
Supplier Article Number:: L09423.09
Glass bottle for solids 2 g ALFAL09423.04
Supplier Article Number:: L09423.04
Expand All / Collapse All

SPECIFICATIONS

Melting point 252-258°
Formula weight 491.37
CAS No. 1253-46-9
Purity 98%
Molecular formula C27H24BrO2P
MDL No. MFCD00082765
Beilstein reference 6535869
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty