(S)-(-)-1-(4-Fluorophenyl)ethyl isocyanate 95%
Supplier: Thermo Scientific
|
Danger
|
ALFAL20231.03EA
290
SGD
ALFAL20231.03
(S)-(-)-1-(4-Fluorophenyl)ethyl isocyanate 95%
(S)-(-)-1-(4-Fluorophenyl)ethyl isocyanate
Formula:
C₉H₈FNO MW: 165,17 g/mol |
MDL Number:
MFCD05664076 CAS Number: 745783-74-8 EINECS: 000-000-0 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...