Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Acetonylpyridinium bromide ≥90%

Supplier: Alfa Aesar

Purity: 90+%
CAS No.: 17282-41-6
Molecular Formula: C8H10BrNO
Formula Weight: 216.08
Melting Point: ca 185°
Beilstein Reference: 3574011


Formula: C₈H₁₀BrNO
CAS Number: 17282-41-6
EINECS: 241-316-0

Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 50 g ALFAB24490.18
Supplier Article Number:: B24490.18
Glass bottle for solids 10 g ALFAB24490.09
Supplier Article Number:: B24490.09
Glass bottle for solids 2 g ALFAB24490.04
Supplier Article Number:: B24490.04
Expand All / Collapse All

SPECIFICATIONS

Melting point ca 185°
Formula weight 216.08
CAS No. 17282-41-6
Purity 90+%
Molecular formula C8H10BrNO
MDL No. MFCD00031948
Beilstein reference 3574011
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty