Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Benzo[b]furan-2,5-diboronic acid ≥98%

Supplier: Alfa Aesar
Warning



Formula: C₈H₈B₂O₅
MW: 205,77
Melting Pt: 293…295 °C
Storage Temperature: Ambient
MDL Number: MFCD08436058
CAS Number: 913835-69-5

 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 250 mg ALFAH53025.MD
Supplier Article Number:: H53025.MD
Expand All / Collapse All

SPECIFICATIONS

Melting point 293-295°
Formula weight 205.77
CAS No. 913835-69-5
Purity 98%
Molecular formula C8H8B2O5
MDL No. MFCD08436058
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty