Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4'-(Trifluoromethylthio)acetophenone ≥97%

Supplier: Alfa Aesar

Purity: 97%
CAS No.: 713-67-7
Molecular Formula: C9H7F3OS
Formula Weight: 220.21
Melting Point: 28-32°
MDL No.: MFCD00236332

Formula: C₉H₇F₃OS
CAS Number: 713-67-7

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 1 g ALFAH26605.03
Supplier Article Number:: H26605.03
Glass bottle for solids 5 g ALFAH26605.06
Supplier Article Number:: H26605.06
Expand All / Collapse All


Melting point 28-32°
Formula weight 220.21
CAS No. 713-67-7
Purity 97%
Molecular formula C9H7F3OS
MDL No. MFCD00236332
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty