Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-(Trifluoromethyl)cyclobutanecarboxylic acid ≥95%

Supplier: Alfa Aesar

Purity: 95%
CAS No.: 277756-45-3
Molecular Formula: C6H7F3O2
Formula Weight: 168.12
Flash Point: 96°(205°F)
Refractive Index: 1.3910
Melting Point: 24-26°
Boiling Point: 76°/14mm
MDL No.: MFCD03425197

 
  Formula: C₆H₇F₃O₂
MDL Number: MFCD03425197
CAS Number: 277756-45-3
Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 500 mg ALFAH25850.ME
Glass bottle for solids 100 mg ALFAH25850.MC
Expand All / Collapse All

SPECIFICATIONS

Melting Point 24-26°
Formula Weight 168.12
CAS No. 277756-45-3
Purity 95%
Refractive Index 1.3910
Flash Point 96°(205°F)
Boiling Point 76°/14mm
Molecular Formula C6H7F3O2
MDL No. MFCD03425197
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty