1-Cyclohexenylboronic acid ≥97%

Warning

Synonyms: Cyclohex-1-en-1-ylboronic acid

ALFAH53247.03EA 723 SGD
ALFAH53247.03
1-Cyclohexenylboronic acid ≥97%
1-Cyclohexenylboronic acid
Formula: C₆H₁₁BO₂
MW: 125,96 g/mol
Melting Pt: 165 °C
MDL Number: MFCD02179497
CAS Number: 89490-05-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR