Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Double bosshead with Allan key and hex-socket set screw

Supplier: Usbeck
Double bosshead made of stainless steel, electrolytical polished.

  • For rods up to 16 mm Ø
  • With allan key and hex-socket set screw

Weight: 111 g



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Material For rod Ø Pk VWR Catalog Number Quantity
Stainless steel 16 mm 1 USBE2206
Supplier Article Number:: 2206
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty