Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Phenyl-1-(trimethylsilyloxy)ethylene ≥97%

Supplier: Alfa Aesar

Purity: 97%
CAS No.: 13735-81-4
Molecular Formula: C11H16OSi
Formula Weight: 192.34
Flash Point: 78°(172°F)
Refractive Index: 1.5020
Boiling Point: 53-54°/1mm
Beilstein Reference: 1306914
MDL No.: MFCD00008582


Formula: C₁₁H₁₆OSi
MDL Number: MFCD00008582
CAS Number: 13735-81-4
EINECS: 237-308-1


 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 1 g ALFAL05308.03
Supplier Article Number:: L05308.03
Glass bottle for solids 5 g ALFAL05308.06
Supplier Article Number:: L05308.06
Expand All / Collapse All

SPECIFICATIONS

Formula weight 192.34
CAS No. 13735-81-4
Purity 97%
Refractive index 1.5020
Flash point 78°(172°F)
Boiling point 53-54°/1mm
Molecular formula C11H16OSi
MDL No. MFCD00008582
Beilstein reference 1306914
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty